1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA113831000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dicyanobutane
Name Note99%
CAS111-69-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger