2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA113580500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1877-71-0
IUPAC Name3-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyWMZNGTSLFSJHMZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point191°C to 197°C
GC>=96.0 %