1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
CAS: 4885-02-3 | C2H4Cl2O | 114.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass bottle
Catalog number FSA113471000
Price (MYR)
1,421.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,421.00
Add to cart
Chemical Identifiers
CAS929-06-6
IUPAC Name2-(2-aminoethoxy)ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyGIAFURWZWWWBQT-UHFFFAOYSA-N
SMILESNCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4570 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid