1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
CAS: 106-46-7 | C6H4Cl2 | 147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number FSA113190025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dichlorobenzene
Name Note97%
CAS106-46-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning