Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
CAS: 74-95-3 | CH2Br2 | 173.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA112831000
Price (MYR)
248.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
248.00
Add to cart
Chemical Identifiers
CAS91464-90-3
IUPAC Name5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
Molecular FormulaC48H80O40
InChI KeyGDSRMADSINPKSL-UHFFFAOYNA-N
SMILESOCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Water=<11 %
Appearance (Color)White to off-white
HPLC>=98.5 %