1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
CAS: 106-93-4 | C2H4Br2 | 187.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA112790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-94-9
IUPAC Name4-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyBHAAPTBBJKJZER-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or granules or flakes
Melting point56°C to 59°C