Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
CAS: 120-46-7 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
Packaging:
View all
Plastic bottle
Catalog number FSA112605000
Price (MYR)
3,112.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,112.00
Add to cart
Chemical Identifiers
CAS570-02-5
IUPAC Name2,4,6-trimethoxybenzoic acid
Molecular FormulaC10H12O5
InChI KeyJATAKEDDMQNPOQ-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C(O)=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Appearance (Color)White to cream