Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
CAS: 120-46-7 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA112600100
Price (MYR)
237.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
237.00
Add to cart
Chemical Identifiers
CAS56601-42-4
IUPAC Nameisothiocyanatocyclopropane
Molecular FormulaC4H5NS
InChI KeyJGFBQFKZKSSODQ-UHFFFAOYSA-N
SMILESS=C=NC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (non-U.S. specification)
Assay from Suppliers CofA≥96.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.5465-1.5525 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow or pale brown