1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Catalog number FSA112470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS36653-82-4
IUPAC Namehexadecan-1-ol
Molecular FormulaC16H34O
InChI KeyBXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or flakes or pellets
GC>=95 %
Appearance (Color)White to off-white