3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 619-05-6 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA112100250
Price (MYR)
397.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
397.00
Add to cart
Chemical Identifiers
CAS122-84-9
IUPAC Name1-(4-methoxyphenyl)propan-2-one
Molecular FormulaC10H12O2
InChI KeyWFWKNGZODAOLEO-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(C)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥97.0%
Refractive Index1.5230-1.5270 @ 20?C
Appearance (Color)Clear colorless to yellow to yellow-green