Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA111845000
Price (MYR)
446.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
446.00
Add to cart
Chemical Identifiers
CAS13967-90-3
IUPAC Namebarium(2+) dibromate
Molecular FormulaBaBr2O6
InChI KeyVEASZGAADGZARC-UHFFFAOYSA-L
SMILES[Ba++].[O-][Br](=O)=O.[O-][Br](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Assay (Iodometric Titration)> 96.0%
Loss on Drying≤ 1.0%
Appearance (Color)White