Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Packaging:
Glass bottle
Catalog number FSA111840010
Price (MYR)
634.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
634.00
Add to cart
Chemical Identifiers
CAS1328-51-4
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
UV spectrumComplies
Appearance (Color)Blue