Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA111630250
Price (MYR)
462.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Chemical Identifiers
CAS2150-44-9
IUPAC Namemethyl 3,5-dihydroxybenzoate
Molecular FormulaC8H8O4
InChI KeyRNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale brown
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)164.0-170.0?C