Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
CAS: 96-41-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA111525000
Price (MYR)
677.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
677.00
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormWire, 0.5mm (0.02in) diameter, annealed.
Assay from Supplier's CofA≥99.95% (metals basis)