Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
CAS: 96-41-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA111521000
Price (MYR)
276.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
276.00
Add to cart
Chemical Identifiers
CAS40876-98-0
IUPAC Namesodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular FormulaC8H11NaO5
InChI KeyJPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES[Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Beige to light orange
Infrared spectrumConforms
Titration with HClO4>=94 %