Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
CAS: 3400-45-1 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA111491000
Price (MYR)
1,898.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,898.00
Add to cart
Chemical Identifiers
CAS27115-49-7
IUPAC Name2-[(3-methylphenyl)formamido]acetic acid
Molecular FormulaC10H11NO3
InChI KeyYKAKNMHEIJUKEX-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)C(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormChunks
Infrared spectrumConforms
Appearance (Color)White
Purity≥96.0%