Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
CAS: 827-52-1 | C12H16 | 160.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA111291000
Price (MYR)
457.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
457.00
Add to cart
Chemical Identifiers
CAS7632-00-0
IUPAC Namesodium nitrite
Molecular FormulaNNaO2
InChI KeyLPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES[Na+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<0.3 %
Titration>=98.5 %
Appearance (Color)Light yellow
Anticaking agent=<0.1 %