Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
CAS: 286-20-4 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA111211000
Price (MYR)
353.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
353.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexene oxide
Name Note98%
CAS286-20-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.