Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA111201000
Price (MYR)
188.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
188.00
Add to cart
Chemical Identifiers
CAS100-69-6
IUPAC Name2-ethenylpyridine
Molecular FormulaC7H7N
InChI KeyKGIGUEBEKRSTEW-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5490 to 1.5510 (20°C, 589 nm)
Stabilizer0.1 % tert.-butylcatechol