Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic Bottle
Catalog number FSA111200025
Price (MYR)
1,087.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,087.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexanone oxime
Melting Point88°C to 92°C
Name Note97%
CAS100-64-1
ColorBeige-Brown to White