1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA111175000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.5 ppm
Copper (Cu)=<0.05 ppm
Cadmium (Cd)=<0.01 ppm
Arsenic (As)=<0.01 ppm
Appearance (Form)Liquid