Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA110890010
Price (MYR)
518.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
518.00
Add to cart
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil
Thickness0.5mm (0.02in)
Total Metal Impurities0.01% max
C-X bond formation reagent
RUO – Research Use Only