4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number FSA110870250
Price (MYR)
225.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
225.00
Add to cart
Chemical Identifiers
CAS62778-12-5
IUPAC Namebis(N1,N1-dimethylbenzene-1,4-diamine); oxalic acid
Molecular FormulaC18H26N4O4
InChI KeyMNUINXKPLPIOEF-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O.CN(C)C1=CC=C(N)C=C1.CN(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)Beige to grey to brown