2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA110851000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Cyanopyridine
Name Note99%
CAS100-70-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning