Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA110650250
Price (MYR)
300.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
300.00
Add to cart
Chemical Identifiers
CAS91-40-7
IUPAC Name2-(phenylamino)benzoic acid
Molecular FormulaC13H11NO2
InChI KeyZWJINEZUASEZBH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point182°C to 187°C
Appearance (Color)Yellow-green to grey-green
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Powder