Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA110630010
Price (MYR)
373.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
373.00
Add to cart
Specifications
Chemical Name or MaterialCumene
Name Note98%
CAS98-82-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.