Coumarin, 99+%, Thermo Scientific Chemicals
Coumarin, 99+%, Thermo Scientific Chemicals
Coumarin, 99+%, Thermo Scientific Chemicals
Coumarin, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Coumarin, 99+%, Thermo Scientific Chemicals
CAS: 91-64-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA110532500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCoumarin
Melting Point68°C to 71°C
Name Note99+%
CAS91-64-5
Health Hazard 1GHS Signal Word: Warning