Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
CAS: 106-22-9 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA110461000
Price (MYR)
289.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
289.00
Add to cart
Chemical Identifiers
CAS69-78-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point240°C to 245°C
Titration with NaOH>=98.5 %
Loss on drying=<0.5 %
Appearance (Color)Off-white to yellow