Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
Packaging:
View all
Fibre drum
Catalog number FSA110450250
Price (MYR)
2,427.00
Add to cart
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (MYR)
2,427.00
Add to cart
Chemical Identifiers
CAS108468-00-4
IUPAC Nametert-butyl N-{[4-(aminomethyl)phenyl]methyl}carbamate
Molecular FormulaC13H20N2O2
InChI KeyNUANLVJLUYWSER-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC1=CC=C(CN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
GC>=89.0 %