Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA110450010
Price (MYR)
494.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Specifications
Chemical Name or MaterialCitric acid
Name NoteAnhydrous
CAS77-92-9
Melting Point153.0°C
Health Hazard 1Warning