Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA110445000
Price (MYR)
909.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
909.00
Add to cart
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.