Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA110441000
Price (MYR)
301.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
301.00
Add to cart
Chemical Identifiers
CAS110-95-2
IUPAC Name[3-(dimethylazaniumyl)propyl]dimethylazanium
Molecular FormulaC7H20N2
InChI KeyDMQSHEKGGUOYJS-UHFFFAOYSA-P
SMILESC[NH+](C)CCC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4225 to 1.4245 (20°C, 589 nm)