Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA110440050
Price (MYR)
218.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
218.00
Add to cart
Chemical Identifiers
CAS28188-41-2
IUPAC Name3-(bromomethyl)benzonitrile
Molecular FormulaC8H6BrN
InChI KeyCVKOOKPNCVYHNY-UHFFFAOYSA-N
SMILESBrCC1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=94 %
Melting point92°C to 97°C