Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA110400250
Price (MYR)
696.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
696.00
Add to cart
Chemical Identifiers
CAS14660-52-7
IUPAC Nameethyl 5-bromopentanoate
Molecular FormulaC7H13BrO2
InChI KeyAFRWBGJRWRHQOV-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4570-1.4610 @ 20?C