Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA110400050
Price (MYR)
343.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Chemical Identifiers
CAS15905-32-5
IUPAC Name3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H8I4O5
InChI KeyOALHHIHQOFIMEF-UHFFFAOYSA-N
SMILESOC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Dye)Dye content: ≥ 95.0%
Loss on Drying≤ 5.0 % (135?C)
Solution TestClear orange solution (0.05% in MeOH)
Appearance (Color)Red to red-orange