trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
CAS: 14371-10-9 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA110350010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS953-26-4
IUPAC Nameethyl (2E)-3-(4-nitrophenyl)prop-2-enoate
Molecular FormulaC11H11NO4
InChI KeyPFBQVGXIMLXCQB-VMPITWQZSA-N
SMILESCCOC(=O)C=CC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)135.0-141.0?C
Appearance (Color)White to pale yellow
Assay (GC)≥98.5%