1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
CAS: 470-82-6 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA110340050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,8-Cineole
CAS470-82-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.