Chrysin, 99+%, Thermo Scientific Chemicals
Chrysin, 99+%, Thermo Scientific Chemicals
Chrysin, 99+%, Thermo Scientific Chemicals
Chrysin, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chrysin, 99+%, Thermo Scientific Chemicals
CAS: 480-40-0 | C15H10O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA110321000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10102-17-7
IUPAC Namedisodium pentahydrate sulfanidesulfonate
Molecular FormulaH10Na2O8S2
InChI KeyPODWXQQNRWNDGD-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Iodimetric>=98.5 %
Total trace metal impurities=<10 ppm