alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
CAS: 104-82-5 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA110171000
Price (MYR)
501.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Chemical Identifiers
CAS1308-96-9
IUPAC Namedieuropium(3+) trioxidandiide
Molecular FormulaEu2O3
InChI KeyAEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Eu+3].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.