Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA110122500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-32-7
IUPAC Name1,3-bis[(9Z)-octadec-9-enoyloxy]propan-2-yl (9Z)-octadec-9-enoate
Molecular FormulaC57H104O6
InChI KeyPHYFQTYBJUILEZ-IUPFWZBJSA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4680 to 1.4700 (20°C, 589 nm)
HPLC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Silylation reagent
RUO – Research Use Only