2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA110011000
Price (MYR)
283.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
283.00
Add to cart
Specifications
Chemical Name or Material2-Chloropyridine
Name Note99%
CAS109-09-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger