4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
CAS: 604-44-4 | C10H7ClO | 178.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA109550250
Price (MYR)
914.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
914.00
Add to cart
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.