4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number FSA109481000
Price (MYR)
283.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
283.00
Add to cart
Chemical Identifiers
CAS1820-80-0
IUPAC Name1H-pyrazol-5-amine
Molecular FormulaC3H5N3
InChI KeyJVVRJMXHNUAPHW-UHFFFAOYSA-N
SMILESNC1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light brown
Appearance (Form)Liquid or fused solid
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
GC>=97.5 %