4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number FSA109480050
Price (MYR)
242.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
242.00
Add to cart
Specifications
Chemical Name or Material4-Chloro-3-methylphenol
Name Note99+%
CAS59-50-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger