1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 2008-75-5 | C7H14ClN·HCl | 184.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA109211000
Price (MYR)
371.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
371.00
Add to cart
Chemical Identifiers
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=95 % (NaOC2H5)
Titration with HCl>=98 % (total NaOC2H5)
NaOH + Na2CO3 (as NaOH)=<2 %