4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 88-04-0 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA109061000
Price (MYR)
216.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
216.00
Add to cart
Specifications
Chemical Name or Material4-Chloro-3, 5-dimethylphenol
Name Note99%
CAS88-04-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning