4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 88-04-0 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA109060025
Price (MYR)
2,118.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,118.00
Add to cart
Chemical Identifiers
CAS52833-63-3
IUPAC Name2-fluoro-N-methylbenzamide
Molecular FormulaC8H8FNO
InChI KeyNAGFMACWWJYORB-UHFFFAOYSA-N
SMILESCNC(=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%