2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
CAS: 78-86-4 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA108830010
Price (MYR)
908.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
908.00
Add to cart
Chemical Identifiers
CAS2530-87-2
IUPAC Name(3-chloropropyl)trimethoxysilane
Molecular FormulaC6H15ClO3Si
InChI KeyOXYZDRAJMHGSMW-UHFFFAOYSA-N
SMILESCO[Si](CCCCl)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4190 to 1.4210 (20°C, 589 nm)