3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA108740050
Price (MYR)
250.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
250.00
Add to cart
Chemical Identifiers
CAS445-29-4
IUPAC Name2-fluorobenzoic acid
Molecular FormulaC7H5FO2
InChI KeyNSTREUWFTAOOKS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% (UK Sourced Material)
Assay (Silylated GC)≥98.0% (UK Sourced Material)
Assay (HPLC)≥98.0% (UK Sourced Material)