4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA108595000
Price (MYR)
585.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Chemical Identifiers
CAS4823-47-6
IUPAC Name2-bromoethyl prop-2-enoate
Molecular FormulaC5H7BrO2
InChI KeyCDZAAIHWZYWBSS-UHFFFAOYSA-N
SMILESBrCCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥92.5%
Comment900-1500 ppm 4-methoxyphenol as stabilizer